Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APRIN Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987957
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7957 Supplier Bioss Supplier No. BS7722R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that interacts with the conserved protein complex termed cohesin. The cohesin complex holds together sister chromatids and facilitates accurate chromosome segregation during mitosis and meiosis. This protein is also a negative regulator of cell proliferation and may be a tumor-suppressor gene.

Specifications

Antigen APRIN
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PDS5B
Gene Accession No. Q9NTI5
Gene Alias AI646570; androgen induced inhibitor of proliferation; Androgen-induced proliferation inhibitor; androgen-induced prostate proliferative shutoff associated protein; androgen-induced prostate proliferative shutoff associated protein AS3; Androgen-induced prostate proliferative shutoff-associated protein AS3; androgen-induced shutoff 3; APRIN; AS3; AW212954; CG008; Kiaa0979; mKIAA0979; PDS5 cohesin associated factor B; PDS5, regulator of cohesion maintenance, homolog B; PDS5, regulator of cohesion maintenance, homolog B (S. cerevisiae); PDS5B; Sister chromatid cohesion protein PDS5 homolog B
Gene Symbols PDS5B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human androgen induced proliferation inhibitor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23047
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.