Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aprotinin from Bovine Lung, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mg
25 mg
Description
General Description
- Aprotinin is a 58-amino acid polypeptide chain found in lung tissue
- Aprotinin can suppress the systemic inflammatory response by reversibly inhibiting various esterases and proteases; it acts as a broad spectrum serine protease inhibitor
Application
- Aprotinin can be used as an enzyme inhibitor in laboratory studies on chymotrypsin, kallikrein, plasmin, and others
- Aprotinin is useful for the laboratory study of post-operative bleeding and wound healing

Specifications
Specifications
For Use With (Application) | Protease Inhibition |
Formulation | Powder |
Quantity | 25 mg |
Source | Bovine Lung |
Storage Requirements | 2° to 8°C |
Regulatory Status | RUO |
Product Type | Aprotinin |
Recombinant | Native |
Form | Lyophilized |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction