Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aprotinin From Bovine Lung
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Description
A useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Not effective against papain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Activity: 5100 to 7300 Kallikrein inhibitor U/mg protein
- Molecular Formula: C284H432N84O79S7
- MDLNumber: MFCD00130541
- CAS: 9087-70-1
Specifications
Specifications
| Concentration | 1 to 2 mg/mL (Soluble in 0.01 M HEPES) |
| For Use With (Application) | Protease Inhibition |
| Molecular Weight (g/mol) | 6511.44 |
| Name | Aprotinin |
| pH Range | 8 |
| Purification Method | Gel filtration |
| Quantity | 10 mg |
| Source | Bovine lung |
| Storage Requirements | 4°C |
| Packaging | Plastic bottle |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction