Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aprotinin From Bovine Lung

Catalog No. aaj11388ma
Click to view available options
Quantity:
10 mg
25 mg
100 mg

Catalog No. AAJ11388MA

Supplier: Thermo Scientific J11388MA

Only null left
Add to Cart

Useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Thermo Scientific™ Aprotinin From Bovine Lung is not effective against papain. | 6511.44 g/mol

A useful protease inhibitor against kallikrein, trypsin, plasmin and chymotrypsin. Not effective against papain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Activity: 5100 to 7300 Kallikrein inhibitor U/mg protein
  • Molecular Formula: C284H432N84O79S7
  • MDLNumber: MFCD00130541
  • CAS: 9087-70-1
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 to 2 mg/mL (Soluble in 0.01 M HEPES)
For Use With (Application) Protease Inhibition
Molecular Weight (g/mol) 6511.44
Name Aprotinin
pH Range 8
Purification Method Gel filtration
Quantity 10 mg
Source Bovine lung
Storage Requirements 4°C
Packaging Plastic bottle
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.