Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aprotinin (From Bovine Lung), Fisher BioReagents

Catalog No. p-24472 Shop All Fisherbrand Products
Click to view available options
Quantity:
10 mL
40 mL
Packaging:
Ampule
This item is not returnable. View return policy
This item is not returnable. View return policy
  • Aprotinin is competitive serine protease inhibitor that inhibits trypsin, chymotrypsin, kallikrein, plasmin, and other proteolytic enzymes
  • Aprotinin is useful in cell and tissue culture studies to prevent protein degradation
  • The aprotinin-protease binding is reversible and will dissociate at extreme pH levels
  • This product is provided as a solution with an activity of ∼10,000 KIU/mL
  •  Intended for research use only
TRUSTED_SUSTAINABILITY

Specifications

Molecular Weight (g/mol) 6511.509
Quantity 10 mL
Source From Bovine Lung
Packaging Ampule
Product Type Aprotinin
Content And Storage 2° to 8°C
Form Powder
Color White
Purity or Quality Grade Analytical
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.