Learn More
Invitrogen™ Aprotinin Monoclonal Antibody (18), Biotin

Description
ABS 037-18B-005 detects Aprotinin from bovine samples. ABS 037-18B-005 has successfully been used in ELISA applications. PBS buffers are unsuitable for this assay and 0.2M glycine sodium buffer, pH 8.8, is recommended. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Specifications
Specifications
| Antigen | Aprotinin |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 18 |
| Concentration | 1.03 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | PTI |
| Gene Accession No. | 0 |
| Gene Alias | aprotinin; basic protease inhibitor; BPI; BPTI; pancreatic trypsin inhibitor; PTI; trypsin ihibitor; trypsin ihibitor precursor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.