Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aprotinin Monoclonal Antibody (18)
GREENER_CHOICE

Catalog No. ABS0371802
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS0371802 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS0371802 Supplier Invitrogen™ Supplier No. ABS0371802
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABS 037-18-02 detects aprotinin in bovine samples. ABS 037-18-02 has been successfully used in ELISA procedures. PBS buffers are unsuitable for these assays and 0.2M glycine sodium buffer, pH 8.8, is recommended. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Aprotinin, also called bovine pancreatic trypsin inhibitor, is a single chain globular Kunitz-type serine proteinase inhibitor found in bovine pancreas, parotid gland and lung. It forms stable, enzymatically inactive complexes with trypsin, chymotrypsin, plasmin and kallikrein. Aprotinin has been used therapeutically as an antifibrinolytic agent under the name Trasylol (Bayer) to reduce blood loss during cardiopulmonary bypass surgery but this use has been associated with an increased frequency of acute renal injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aprotinin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 18
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene PTI
Gene Accession No. 0
Gene Alias aprotinin; basic protease inhibitor; BPI; BPTI; pancreatic trypsin inhibitor; PTI; trypsin ihibitor; trypsin ihibitor precursor
Gene Symbols PTI
Host Species Mouse
Immunogen Aprotinin from bovine lung.
Purification Method Protein A/G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404172
Target Species Bovine
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.