Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APRT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535305
Description
APRT Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Adenine phosphoribosyltransferase belongs to the purine/pyrimidine phosphoribosyltransferase family. A conserved feature of this gene is the distribution of CpG dinucleotides. This enzyme catalyzes the formation of AMP and inorganic pyrophosphate from adenine and 5-phosphoribosyl-1-pyrophosphate (PRPP). It also produces adenine as a by-product of the polyamine biosynthesis pathway. A homozygous deficiency in this enzyme causes 2,8-dihydroxyadenine urolithiasis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
APRT | |
Polyclonal | |
Unconjugated | |
Aprt | |
adenine phosphoribosyl transferase; adenine phosphoribosyltransferase; AMP; AMP diphosphorylase; AMP pyrophosphorylase; APRT; APRTD; C85684; DKFZp686D13177; MGC125856; MGC125857; MGC129961; transphosphoribosidase | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
353 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P07741 | |
Aprt | |
KLH conjugated synthetic peptide between 12-40 amino acids from the N-terminal region of human APRT. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction