Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APRT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596330 Supplier Invitrogen™ Supplier No. PA596330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADSELQLVE QRIRSFPDFP TPGVVFRDIS PVLKDPASFR AAIGLLARHL KATHGGRIDY IAGLDSRGFL FGPSLAQELG LGCVLIRKRG KLPGPTLWAS YSLEYGKAEL EIQKDALEPG QRVVVVDDLL ATGGTMNAAC ELLGRLQAEV LECVSLVELT SLKGREKLAP VPFFSLLQYE; Positive Samples: MCF7, 22Rv1, THP-1, A-549, K-562, HeLa, Mouse intestine; Cellular Location: Cytoplasm.

Adenine phosphoribosyltransferase belongs to the purine/pyrimidine phosphoribosyltransferase family. A conserved feature of this gene is the distribution of CpG dinucleotides. This enzyme catalyzes the formation of AMP and inorganic pyrophosphate from adenine and 5-phosphoribosyl-1-pyrophosphate (PRPP). It also produces adenine as a by-product of the polyamine biosynthesis pathway. A homozygous deficiency in this enzyme causes 2,8-dihydroxyadenine urolithiasis. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APRT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Aprt
Gene Accession No. P07741, P08030
Gene Alias adenine phosphoribosyl transferase; adenine phosphoribosyltransferase; AMP; AMP diphosphorylase; AMP pyrophosphorylase; APRT; APRTD; C85684; DKFZp686D13177; MGC125856; MGC125857; MGC129961; transphosphoribosidase
Gene Symbols Aprt
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human APRT (NP_0004761).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11821, 353
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.