Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APS Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984925
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4925 Supplier Bioss Supplier No. BS2731R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

APS (adapter protein with Pleckstrin homology and Src homology 2 domains) a member of the Lnk family, is an adaptor protein that is involved in B-cell signaling, insulin signaling, cytoskeletal reorganization. It is tyrosine phosphorylated by JAK2, KIT and other kinases during B-cell receptor stimulation of many different cytokines, chemokines and leukokines. APS has been shown to inhibit the JAK-STAT pathway in collaboration with c-Cbl. It is located in the cytoplasm in pre-PDGF (platelet derived growth factor) stimulated cells and localizes to the plasma membrane and peripheral regions upon PDGF stimulation. This protein is expressed in B-cells and mast cells when detected in haematopoietic cell lines yet is also expressed in many different tissues including spleen, prostrate, testis, uterus, small intestine and skeletal muscle.

Specifications

Antigen APS
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SH2B2
Gene Accession No. O14492
Gene Alias Adapter protein with pleckstrin homology and Src homology 2 domains; Alpha-1-antitrypsin; Aps; SH2 and PH domain-containing adapter protein APS; SH2B adapter protein 2; SH2B adaptor protein 2; Sh2b2
Gene Symbols SH2B2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human APS.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10603
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.