Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AQ-RA 741
GREENER_CHOICESDP

Catalog No. 229250 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-925-0 50 mg
22-921-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-925-0 Supplier R&D Systems™ Supplier No. 2292/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective, high affinity M2 antagonist

High affinity, selective muscarinic M2 receptor antagonist (pKi values are 8.3, 7.7 and 6.82 for M2, M1 and M3 receptors, respectively). Displays cardioselectivity in vivo, over intestinal, tracheal and bladder muscarinic receptors; inhibits vagally and agonist-induced bradycardia.

Chemical Identifiers

CAS 123548-16-3
Molecular Formula C27H37N5O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 463.62
Purity >99%
Molecular Weight (g/mol) 463.626
Chemical Name or Material AQ-RA 741
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.