Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AQP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983423
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3423 Supplier Bioss Supplier No. BS1506R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aquaporin 1 is a 28kD integral membrane protein which was originally identified in red blood cells and renal proximal tubules. AQP1 is also expressed by the choroid plexus and various other tissues. It forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.

Specifications

Antigen AQP1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Aqp1
Gene Accession No. P29972
Gene Alias AQP 1; AQP CHIP; AQP1; AQP-1; AQP-CHIP; aquaporin 1; Aquaporin 1 (aquaporin channel forming integral protein 28 (CHIP)); aquaporin 1 (channel-forming integral protein, 28kDa, CO blood group); aquaporin 1 (Colton blood group); aquaporin 1, Colton blood group antigen; Aquaporin CHIP; Aquaporin1; Aquaporin-1; Aquaporin-CHIP; channel-like integral membrane protein, 28-kDa; CHIP 28; Chip28; CO; Colton blood group; Delayed early response protein 2; DER2; growth factor-induced delayed early response protein; MGC26324; Urine water channel; Water channel protein for red blood cells and kidney proximal tubule
Gene Symbols Aqp1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AQP1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 358
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.