Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AQP12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548287
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548287 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548287 Supplier Invitrogen™ Supplier No. PA548287
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AQP12B Polyclonal Antibody for Western Blot, IHC (P), Flow

Aquaporins facilitate the transport of water and small neutral solutes across cell membranes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AQP12B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene AQP12B
Gene Accession No. A6NM10
Gene Alias AQP12B; AQP-12B; aquaporin 12B; aquaporin-12B; INSSA3; insulin synthesis associated 3
Gene Symbols AQP12B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 236-264 amino acids from the C-terminal region of human AQP12B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653437
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.