Learn More
AQP7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 251311AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-4739
Only null left
Add to Cart
Description
Description
Aquaporins/major intrinsic protein (MIP) are a family of water-selective membrane channels. Aquaporin 7 has greater sequence similarity with AQP3 and AQP9 and they may be a subfamily. Aquaporin 7 and AQP3 are at the same chromosomal location suggesting that 9p13 may be a site of an aquaporin cluster. Aquaporin 7 facilitates water, glycerol and urea transport. It may play an important role in sperm function.
Specifications
Specifications
AQP7
Polyclonal
Unconjugated
AQP7
AQP 7, AQP7, AQP7L, AQP9, AQPap, Aquaglyceroporin 7, aquaporin 7, Aquaporin 7 like, Aquaporin adipose
Rabbit
Antigen Affinity Chromatography
RUO
11832, 364
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14520, O54794
AQP7
AQP7 Fusion Protein Ag17945
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.