Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AQPEP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562765 Supplier Invitrogen™ Supplier No. PA562765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPYPIKDVVL CYGIALGSDK EWDILLNTYT NTTNKEEKIQ LAYAMSCSKD PWILNRYMEY AISTSPFTSN ETNIIEVVAS SEVGRYVA Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 61%.

Metalloprotease which may be important for placentation by regulating biological activity of key peptides at the embryo-maternal interface. On synthetic substrates it shows a marked preference for Leu-4-methylcoumaryl-7-amide (Leu-MCA) over Met-MCA, Arg-LCA and Lys-LCA. Cleaves the N-terminal amino acid of several peptides such as angiotensin-3, kisspeptin-10 and endokinin C. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AQPEP
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LVRN
Gene Accession No. Q6Q4G3
Gene Alias Aminopeptidase Q; APQ; AP-Q; AQPEP; CHL2 antigen; laeverin; LVRN; TAQPEP
Gene Symbols LVRN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AQPEP. Recombinant protein control fragment (Product #RP-100461).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 206338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.