Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AQR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109403 Supplier Invitrogen™ Supplier No. PA5109403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEFLLELLVS RHERRISQIQ QLNQMPLYPT EKIIWDENIV PTEYYSGEGC LALPKLNLQF LTLHDYLLRN FNLFRLESTY EIRQDIEDSV SRMKPWQSEY GGVVFGGWAR MAQPIVAFTV VEVAKPNIGE NWPTRVRADV TINLNVRDHI KDEWEGLRKH DVCFLITVRP TKPYGTKFDR RRPFIEQVGL VYVRGCEIQG MLDDKGRVIE DGPEPRPNLR GESRTFRVFL DPNQYQQDMT NTIQNGAEDV YETFNIIMRR KPKENNFKAV LETIRNLMNT DCVVPDWLHD IILGYGDPSS A.

Involved in pre-mRNA splicing as component of the spliceosome (PubMed:11991638, PubMed:25599396, PubMed:28502770, PubMed:28076346). Intron-binding spliceosomal protein required to link pre-mRNA splicing and snoRNP (small nucleolar ribonucleoprotein) biogenesis (PubMed:16949364). Plays a key role in position-dependent assembly of intron-encoded box C/D small snoRNP, splicing being required for snoRNP assembly (PubMed:16949364). May act by helping the folding of the snoRNA sequence. Binds to intron of pre-mRNAs in a sequence-independent manner, contacting the region between snoRNA and the branchpoint of introns (40 nucleotides upstream of the branchpoint) during the late stages of splicing (PubMed:16949364). Has ATP-dependent RNA helicase activity and can unwind double-stranded RNA molecules with a 3' overhang (in vitro) (PubMed:25599396). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AQR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.95 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AQR
Gene Accession No. O60306, Q8CFQ3
Gene Alias Aqr; aquarius; aquarius homolog; aquarius intron-binding spliceosomal factor; AW495846; fSAP164; functional spliceosome-associated protein 164; IBP160; intron-binding protein aquarius; intron-binding protein of 160 kDa; Kiaa0560; mKIAA0560; RNA helicase aquarius
Gene Symbols AQR
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-700 of human AQR (NP_055506.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11834, 366163, 9716
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.