Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AquaBath™ Water Baths

Catalog No. p-4527593
Click to view available options
Capacity (Metric):
20 L
Controller Type:
Analog
Digital
Voltage:
230 V
Frequency:
50/60 Hz
This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Catalog No. 15-476-14Q

Supplier: Thermo Scientific™ 18100A1CEQ

This item is not returnable. View return policy
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.

Aquabath Water Baths provide precise temperature control and are available with an analog or digital operating system, in a variety of sizes from 2 to 28L.

Thermo Scientific™ Aquabath Water Baths provide precise temperature control and are available with an analog or digital operating system. The Aquabath is available in a variety of sizes from 2 to 28L, including a shallow form to meet specific application needs and budgets. Bath features dual thermostats and an independent high-limit thermostat for overtemperature protection.

  • Hinged acrylic cover lifts to a 90° stay-open position or can be removed completely
  • “Fins” on the hinged clear acrylic gable cover (included) protect hands from hot vapors
  • Exterior remains cool to the touch even after extended use to prevent accidental burns
  • Temperature-resistant plastic rim fits over the seamless stainless-steel reservoir for a leak-free seal
  • Stainless-steel exterior resists corrosion and has a tough, easy-to-clean enamel coating
  • Rounded, seamless stainless-steel reservoir and diffuser tray resist rust and contamination for simple maintenance
  • Movable temperature controllers on the 20 and 28L digital models can be used on the bath long or short side to best utilize bench space
  • Heating elements will not burn out if the bath accidentally runs dry
  • Drain pump is included with 10, 20, 28 and 5/10L models

Digital Operating System

  • Field-calibratable
  • PID control provides ±0.24°C uniformity, ±0.5°C stability and ±0.1°C control resolution at 37°C
  • Digital temperature set and readout selectable display with 0.1° resolution

Analog Operating System

  • Easy-to-use analog control and hydraulic thermostat—ideal for fixed-setpoint applications
  • Temperature uniformity ±0.2°C at 37°C
  • Diffuser tray, thermometer clip and nonhazardous liquid, partial immersion (76mm) thermometer (-10° to +110°C) included (230V models do not include a thermometer)

Ordering Information:

Stainless-steel covers available separately.

Compatible with:

  • Cover, for 2L baths, stainless steel (19000-11Q)
  • Cover, for 2L shallow and 5L baths, stainless steel (19000-13Q)
  • Cover, for 20L and 28L baths, stainless steel (19000-17Q)
  • Cover, for 10L baths, stainless steel (19000-15Q)
TRUSTED_SUSTAINABILITY

Specifications

Chamber Material Stainless Steel
Controller Type Analog
Capacity (Metric) 20 L
Certifications/Compliance CSA and CE certified
Pump Type Drain Pump
Temperature Range Ambient to 100°C
Material Enamel-coated Stainless Steel Shell
Temperature Uniformity ±0.2 at 37°C
Dimensions (L x W x H) 16.4 x 24.2 x 7.6 in. (42 x 62 x 19 cm)
Length (English) 16.4 in.
Length (English) Interior 11.8 in.
Length (Metric) 42 cm
Length (Metric) Interior 30 cm
Width (English) 24.2 in.
Width (English) Interior 19.8 in.
Width (Metric) 62 cm
Width (Metric) Interior 50 cm
Height (English) 7.6 in.
Height (Metric) 19 cm
Depth (English) Interior 6 in.
Depth (Metric) Interior 15 cm
Shipping Weight (English) 22.3 lb.
Shipping Weight (Metric) 10.1 kg
Voltage 230 V
Frequency 50/60 Hz
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.