Learn More
Thermo Scientific™ Orion™ AQUAfast™ Reaction/Digestion Tubes and Liquid Reagents
Catalog No. 13301162
Change view
Click to view available options
Type:
COD
COD High Range
COD Low Range
COD Mid Range
Range:
0 to 150 mg/L O2
0 to 1500 mg/L O2
0 to 15000 mg/L O2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Range
13-301-162 COD Mid Range 0 to 1500 mg/L O2
13-642-043 COD 0 to 1500 mg/L O2
13-642-044 COD 0 to 15000 mg/L O2
13-642-042 COD 0 to 150 mg/L O2
13-301-163 COD High Range 0 to 15000 mg/L O2
13-301-159 COD Low Range 0 to 150 mg/L O2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 13-301-162 Supplier Thermo Scientific™ Supplier No. CODM150
Only null left
Add to Cart
Add to Cart
Perform colorimetric testing including fluoride, COD, total nitrogen and total phosphate using the Thermo Scientific™ Orion™ AQUAfast Liquid Reagents, Digestion and Reaction Tubes.
Use Thermo Scientific™ Orion™ AQUAfast Digestion Tubes, Reaction Tubes and Liquid Reagents with Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. Digestion and reaction tubes contain the precise amount of reagent, are made of special optical glass and fit the Orion™ AQUAfast COD165 thermoreactor. Digestion tubes measure COD, total nitrogen and total phosphate. Reaction tubes measure ammonia, nitrate and phosphate. Liquid reagents mix quickly and easily in solution and measure fluoride (SPADN), hydrazine and pH.
Changes in nutrient levels can alter the delicate balance in ecosystems and contaminate drinking water. Tube tests make it easy to determine concentrations of species in samples.
TRUSTED_SUSTAINABILITY
Specifications
Description AQUAfast COD 0-1500 ppm digestion tubes
For Use With (Equipment) Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast
Includes 150 Mid Range COD Digestion Tubes
Quantity 150 Tubes
Range 0 to 1500 mg/L O2
Test Method Dichromate/H2SO4
Type COD Mid Range
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.