Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ AquaPhenol™

Catalog No. p-8010843
Click to view available options
Quantity:
250 mL

Water saturated phenol for DNA/RNA extractions

Phenol is used as a reagent in chemical analysis. It is used in the manufacture of colorless or light-colored artificial resins, many medical and industrial organic compounds and dyes. It is commonly used in tissue preservation and DNA/RNA extraction with chloroform.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at +4°C. Store under nitrogen. Protect from light.
Formulation Water saturated Phenol
pH 4.5 - 5 (Lit.)
Research Category Molecular Biology
Quantity 250 mL
Form Solution
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.