Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aquaporin 0 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114870
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114870 Supplier Invitrogen™ Supplier No. PA5114870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AQP0.

AQP0 is the most abundant endogenous protein in the plasma membrane of lens fiber cells where it functions not only as a water pore, but it is also involved in fiber-fiber adhesion and is crucial for fiber cell structure and organization. AQP0 contains an additional pore constriction, not seen in any other aquaporin structures, which may be responsible for pore gating. The closed AQP0 pore holds just three water molecules, which are spaced too far apart to form hydrogen bonds with each other. The C-terminal domain of AQP0 undergoes extensive post-translational modification, including many truncations, during lens aging due to the actions of m-calpain, proteases or non-enzymatic mechanisms. These truncation sites may be involved in the development of cataracts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 0
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MIP
Gene Accession No. P09011, P30301, P51180
Gene Alias AQP0; aquaporin 0; Aquaporin0; aquaporin-0; Cat; CTRCT15; dominant cataract; Hfi; hydropic fibers; lens fiber major intrinsic protein; lens opacity; LIM1; Lop; major intrinsic protein of eye lens fiber; major intrinsic protein of lens fiber; MIP; MIP26; MP26; Palm; shrivelled; Svl
Gene Symbols MIP
Host Species Rabbit
Immunogen A synthesized peptide derived from human MIP(Accession P30301), corresponding to amino acid residues V125-G175.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17339, 25480, 4284
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.