Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin 1/AQP1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299068MFV610
Description
Aquaporin 1/AQP1 Polyclonal antibody specifically detects Aquaporin 1/AQP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Aquaporin 1/AQP1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
28-kDa, AQP-1, AQP-CHIP, aquaporin 1 (channel-forming integral protein, 28kDa), aquaporin 1 (Colton blood group), Aquaporin-CHIP, CHIP2828kDa, CO blood group), CO, MGC26324 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Aquaporin 1/AQP1. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
358 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction