Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin 1/AQP1 Antibody (OTI2D10), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270199MFV500
Description
Aquaporin 1/AQP1 Monoclonal antibody specifically detects Aquaporin 1/AQP1 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
Aquaporin 1/AQP1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Flow Cytometry | |
OTI2D10 | |
Flow Cytometry | |
28-kDa, AQP-1, AQP-CHIP, aquaporin 1 (channel-forming integral protein, 28kDa), aquaporin 1 (Colton blood group), Aquaporin-CHIP, CHIP2828kDa, CO blood group), CO, MGC26324 | |
Full length human recombinant protein of human AQP1 (NP_932766) produced in HEK293T cell. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
358 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction