Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aquaporin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577842
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577842 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577842 Supplier Invitrogen™ Supplier No. PA577842
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

0.5 μg/mL of PA5-77842 was sufficient for detection of aquaporin 1 in 10 μg of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 28.5kDa. May detect larger glycosylated bands approximately 35-50kDa.

Aquaporin 1 is a 28kD integral membrane protein which was originally identified in red blood cells and renal proximal tubules. AQP1 is also expressed by the choroid plexus and various other tissues. It forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 1
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Aqp1
Gene Accession No. P29972, P29975, Q02013
Gene Alias AQP 1; AQP CHIP; Aqp1; AQP-1; AQP-CHIP; aquaporin 1; Aquaporin 1 (aquaporin channel forming integral protein 28 (CHIP)); aquaporin 1 (channel-forming integral protein, 28kDa, CO blood group); aquaporin 1 (Colton blood group); aquaporin 1, Colton blood group antigen; Aquaporin CHIP; Aquaporin1; aquaporin-1; Aquaporin-CHIP; channel-like integral membrane protein, 28-kDa; CHIP 28; CHIP28; CO; Colton blood group; Delayed early response protein 2; DER2; growth factor-induced delayed early response protein; membrane channel; MGC26324; urine water channel; Water channel protein for red blood cells and kidney proximal tubule
Gene Symbols Aqp1
Host Species Rabbit
Immunogen Produced against the N-terminal peptide (sequence N-MASEFKKKLF) of rat Aquaporin 1 (aa 1-10).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11826, 25240, 358
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.