Learn More
Invitrogen™ Aquaporin 11 Polyclonal Antibody

Description
Aquaporin 11 Polyclonal Antibody for Flow, IHC (P), Western Blot
Specifications
Specifications
| Antigen | Aquaporin 11 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | AQP11 |
| Gene Accession No. | Q8BHH1, Q8NBQ7 |
| Gene Alias | 1700015P13Rik; AI648923; AQP11; AQP-11; AQPX1; aquaporin 11; aquaporin-11; CG12251 gene product; PSEC0027; sjds |
| Gene Symbols | AQP11 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.