Learn More
Invitrogen™ Aquaporin 12 Polyclonal Antibody

Description
Antibody detects endogenous levels of total AQP12.
Specifications
Specifications
| Antigen | Aquaporin 12 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | AQP12B |
| Gene Accession No. | A6NM10, Q8IXF9 |
| Gene Alias | AQP12; AQP-12; AQP12A; AQP12B; AQP-12B; AQPX2; aquaporin 12; aquaporin 12A; aquaporin 12B; aquaporin X2; aquaporin-12; aquaporin-12A; Aquaporin-12B; INSSA3; insulin synthesis associated 3; RGD1562727 |
| Gene Symbols | AQP12A, AQP12B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.