Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-12A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP280501
Description
Aquaporin-12A Polyclonal specifically detects Aquaporin-12A in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Aquaporin-12A | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunohistochemistry-Paraffin 1:1000 | |
AQP-12, AQP12AQPX2aquaporin X2, aquaporin 12, aquaporin 12A, aquaporin-12A | |
Rabbit | |
Unpurified | |
RUO | |
375318 | |
Human, Mouse, Monkey |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Lyophilized from whole serum | |
AQP12A | |
A synthetic peptide from amino acid region 10-50 of human Aquaporin-12A conjugated to blue carrier protein was used as the antigen. | |
0.1 mL | |
Primary | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction