Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Aquaporin-2 Antibody Blocking Peptide

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 359 |
Concentration | 1 mg/mL |
Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB110-74682 |
Gene Alias | ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD |
Gene Symbol | AQP2 |
Quantity | 0.1 mg |
Research Category | Phospho Specific, Plasma Membrane Markers, Signal Transduction |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction