Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 26 publications
$268.00 - $489.50
Specifications
Antigen | Aquaporin-2 |
---|---|
Concentration | 1 mg/ml |
Dilution | Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1 ug/ml, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen reported in scientific literature |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11074682T
![]() |
Novus Biologicals
NB11074682SS |
0.025 mL |
Each for $268.00
|
|
|||||
NB11074682
![]() |
Novus Biologicals
NB11074682 |
0.1 mL |
Each for $489.50
|
|
|||||
Description
Aquaporin-2 Polyclonal specifically detects Aquaporin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Aquaporin-2 | |
Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1 ug/ml, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen reported in scientific literature | |
Polyclonal | |
Rabbit | |
Plasma Membrane Markers, Signal Transduction | |
PBS with 0.05% Sodium Azide | |
ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD | |
AQP2 | |
IgG | |
Affinity Purified | |
Human, rat and mouse. 100% sequence identity with sheep, porcine, and bovine proteins. |
1 mg/ml | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
P34080 | |
359 | |
Synthetic peptide made to a C-terminus portion of rat Aquaporin-2 (within residues 200-300). [Swiss-Prot# P34080] | |
Primary | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title