Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11074682MFV450
Description
Aquaporin-2 Polyclonal antibody specifically detects Aquaporin-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
Aquaporin-2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD | |
Synthetic peptide made to a C-terminus portion of rat Aquaporin-2 (within residues 200-300). [Swiss-Prot# P34080] | |
0.1 mL | |
Phospho Specific, Plasma Membrane Markers, Signal Transduction | |
359 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction