Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-2, p Ser261 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB100611000.02ML
Description
Aquaporin-2 Polyclonal specifically detects Aquaporin-2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot.Specifications
Aquaporin-2 (p Ser261) | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100, Dot Blot | |
P34080 | |
AQP2 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser261 conjugated to KLH | |
Affinity Purified | |
RUO | |
Primary | |
Specific for ∼29k AQP2 protein phosphorylated at Ser261. Also recognizes the glycosylated form of AQP2 at ∼ 37k. Immunolabeling of the AQP2 band is blocked by preadsorption with the phospho-peptide used as antigen but not by the corresponding dephospho-peptide. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD | |
Rabbit | |
29 kDa | |
0.02 mL | |
Phospho Specific | |
359 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction