Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-2, p Ser261 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB10061100
Description
Aquaporin-2 Polyclonal specifically detects Aquaporin-2 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Dot Blot.Specifications
Aquaporin-2 (p Ser261) | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:100 Immunohistochemistry-Paraffin 1:100 Dot Blot | |
P34080 | |
AQP2 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser261 conjugated to KLH. Accession # P34080 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for ∼29k AQP2 protein phosphorylated at Ser261. Also recognizes the glycosylated form of AQP2 at ∼ 37k. Immunolabeling of the AQP2 band is blocked by preadsorption with the phospho-peptide used as antigen but not by the corresponding dephospho-peptide. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Dot Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
ADH water channel, AQP-2, AQP-CD, aquaporin 2 (collecting duct), aquaporin-2, aquaporin-CD, Collecting duct water channel protein, MGC34501, Water channel protein for renal collecting duct, water-channel aquaporin 2, WCH-CD | |
Rabbit | |
29 kDa | |
0.1 mL | |
Phospho Specific | |
359 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction