Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin-3 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen Aquaporin-3
Concentration LYOPH
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018703
SDP
View Documents
Novus Biologicals
NBP1703760.025ML
0.025 mL
Each for $181.90
Only null left
Add to Cart
 
NBP170376
SDP
View Documents
Novus Biologicals
NBP170376
0.1 mL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Aquaporin-3 Polyclonal specifically detects Aquaporin-3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Aquaporin-3
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Goat
Membrane Trafficking and Chaperones
AQP-3, Aquaglyceroporin-3, aquaporin 3, aquaporin 3 (GIL blood group), aquaporin 3 (Gill blood group), aquaporin-3, GIL, Gill blood group
AQP3
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
LYOPH
Polyclonal
Antisera
RUO
Mouse, Rat
360
IgG
Unpurified
Rat, mouse. Other species not yet tested.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.