Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aquaporin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119892 Supplier Invitrogen™ Supplier No. PA5119892
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Mouse kidney tissue lysates, human esophagus tissue, human kidney tissue, human prostate carcinoma tissue, rat bladder tissue, rat stomach tissue. Subcellular Location: Cell membrane, Basolateral cell membrane.

AQP3 is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 3 is localized at the basal lateral membranes of collecting duct cells in the kidney. In addition to its water channel function, aquaporin 3 has been found to facilitate the transport of nonionic small solutes such as urea and glycerol, but to a smaller degree. It has been suggested that water channels can be functionally heterogeneous and possess water and solute permeation mechanisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene AQP3
Gene Accession No. P47862, Q8R2N1, Q92482
Gene Alias 31.4 kDa water channel protein; AQP 3; AQP-2; AQP3; AQP-3; Aquaglyceroporin-3; aquaporin 3; aquaporin 3 (GIL blood group); aquaporin 3 (Gill blood group); Aquaporin3; aquaporin-3; GIL; Gill blood group
Gene Symbols AQP3
Host Species Rabbit
Immunogen Synthetic peptide within human Aquaporin 3 aa 243-292.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11828, 360, 65133
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.