Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-4 Antibody (22F10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Aquaporin-4 |
---|---|
Clone | 22F10 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Aquaporin-4 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
aquaporin 4, MGC22454 | |
A synthesized peptide derived from Human Aquaporin-4 [UniProt P55087] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
22F10 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
361 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title