Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aquaporin 4 Monoclonal Antibody (C9)
Mouse Monoclonal Antibody
$213.50 - $522.50
Specifications
Antigen | Aquaporin 4 |
---|---|
Clone | C9 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541556
|
Invitrogen™
MA541556 |
200 μL |
Each for $522.50
|
|
|||||
PIMA543952
|
Invitrogen™
MA543952 |
20 μL |
Each for $213.50
|
|
|||||
Description
Aquaporin 4 Monoclonal Antibody for Western Blot, IHC (P)
In skeletal muscle, AQP4 (aquaporin 4 also known as mercurial insensitive water channel), localizes to the sarcolemma of fast-twitch muscle fibers. Aquaporins (AQPs) are a large family of integral membrane water transport channel proteins that facilitate the transport of water through the cell membrane. This function is conserved in animals, plants and bacteria. Many isoforms of aquaporin have been identified in mammals, designated AQP0 through AQP10. Aquaporins are widely distributed and it is not uncommon for more than one type of AQP to be present in the same cell. Although most aquaporins are only permeable to water, AQP3, AQP7, AQP9 and one of the two AQP10 transcripts are also permeable to urea and glycerol.Specifications
Aquaporin 4 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P55087 | |
361 | |
Recombinant protein Aquaporin 4. The antigen corresponds to amino acid range 178-317 of the target protein. | |
Antibody |
C9 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
AQP4 | |
Aqp4; AQP-4; aquaporin 4; aquaporin type4; Aquaporin4; aquaporin-4; HMIWC2; Mercurial-insensitive water channel; MGC22454; MIWC; MIWC2; WCH4 | |
AQP4 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title