Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin-4 Mouse anti-Human, Clone: AQP4/3324, Novus Biologicals™
SDP

Catalog No. p-7244828 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177861 20 μg
NB177860 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177861 Supplier Novus Biologicals Supplier No. NBP31395220UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Aquaporin-4
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone AQP4/3324
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P55087
Gene Alias aquaporin 4, MGC22454
Host Species Mouse
Immunogen Human recombinant Aquaporin-4 protein fragment (around aa254-323) (exact sequence is proprietary) (Uniprot: P55087)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 361
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.