Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin-4, Mouse anti-Rat, Human, Mouse, Clone: mECD, Millipore Sigma™

Catalog No. MABN2471100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2471100 100 μg
MABN247125 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2471100 Supplier MilliporeSigma Supplier No. MABN2471100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Aquaporin-4, clone mECD, Cat. No. MABN2471, is a highly specific mouse monoclonal antibody that targets Aquaporin-4 and has been tested for use in Flow Cytometry, Immunohistochemistry, Immunofluorescence, and Immunoprecipitation.

Aquaporin-4 (UniProt: P55087; also known as AQP-4, Mercurial-insensitive water channel, MIWC, WCH4) is encoded by the AGP4 gene (Gene ID: 361) in human. Aquaporin-4 is a multi-pass membrane protein that forms a water-specific channel and serves as an osmoreceptor that regulates body water balance and mediates water flow within the central nervous system. Its extracellular domains are localized to amino acids 58-64, 137-155, and 206-231); its transmembrane domains are in amino acids 37-57, 65-85, 116-136, 156-176, 185-205, and 232-252); and the cytoplasmic domain is in amino acids 1-36, 86-115, 177-184, and 253-323). Aquaporin-4 is largely expressed in the brain and the skeletal muscle and at lower levels in heart, kidney, and lung tissue. Phosphorylation of Aquaporin-4 at Serine 180 by protein kinase C is shown to reduce conductance by about 50%, whereas phosphorylation at Serine 111 by protein kinase G in response to glutamate can increase conductance by about 40%. Two isoforms of Aquaporin-4 have been described that are produced by alternative splicing. Antibodies to Aquaporin-4 can serve as useful serum markers to distinguishing neuromyelitis optica (NMO) from multiple sclerosis. Clone mECD is shown to induce clustering and internalization of Aquporin-4 and cause its redistribution, which is also observed with NMO-IgG. Exposure to mECD antibody is also shown to cause coaggregation of astrocytic CD28B and Aquaporin-4. (Ref.: Hinson, SR et al. (2017). Proc. Natl.Acad. Sci. USA 114(21); 5491-5496).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin-4
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone mECD
Conjugate Unconjugated
Dilution Flow Cytometry Analysis: A representative lot detected Aquaporin-4 in Flow Cytometry applications (Hinson, S.R., et. al. (2017). Proc Natl Acad Sci USA. 114(21):5491-5496).Immunofluorescence Analysis: A representative lot detected Aquaporin-4 in Immunofluorescence applications (Hinson, S.R., et. al. (2017). Proc Natl Acad Sci USA. 114(21):5491-5496).Immunoprecipitation Analysis: A representative lot immunoprecipitated Aquaporin-4 in Immunoprcipitation applications (Hinson, S.R., et. al. (2017). Proc Natl Acad Sci USA. 114(21):5491-5496).
Formulation Purified mouse monoclonal antibody IgG1 in PBS without preservatives.
Gene Alias AQP-4;Mercurial-insensitive water channel;MIWC;WCH4
Gene Symbols AQP4
Host Species Mouse
Immunogen His-tagged full-length human recombinant Aquaporin-4.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001641
Test Specificity Clone mECD specifically detects Aquaporin-4 in human, mouse, and rat brain tissue sections. It targets an epitope with in the extracellular region.
Target Species Rat, Human, Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.