Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin 4, Rabbit anti-Rat, Mouse, Human, Polyclonal, Millipore Sigma™

Catalog No. ABN91025UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN91025UG 25 μg
ABN910100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN91025UG Supplier MilliporeSigma Supplier No. ABN91025UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Aquaporin 4, Cat. No. ABN910, is a highly specific rabbit polyclonal antibody that targets Aquaporin 4 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Aquaporin-4 (UniProt: P47863; also known as AQP-4, Mercurial-insensitive water channel, MIWC) is encoded by the Aqp4 gene (Gene ID: 25293) in rat. Aquaporin-4 is a multi-pass membrane homotetrameric protein that forms a water-specific channel and serves as an osmoreceptor that regulates body water balance and mediates water flow within the central nervous system. Its extracellular domains are localized to amino acids 58-64, 137-155, and 206-231); its transmembrane domains are in amino acids 37-57, 65-85, 116-136, 156-176, 185-205, and 232-252); and the cytoplasmic domain is in amino acids 1-36, 86-115, 177-184, and 253-323). Aquaporin-4 is largely expressed in the brain and the skeletal muscle and at lower levels in heart, kidney, and lung tissue. Phosphorylation of Aquaporin-4 at Serine 180 by protein kinase C is shown to reduce conductance by about 50%, whereas phosphorylation at Serine 111 by protein kinase G in response to glutamate can increase conductance by about 40%. Two isoforms of Aquaporin-4 have been described that are produced by alternative splicing. Antibodies to Aquaporin-4 can serve as useful serum markers to distinguishing neuromyelitis optica (NM) from multiple sclerosis. Clone mCCD does not cause redistribution of Aquaporin-4 and also it does not affect coaggregation astrocytic CD32-B and Aquaporin-4. (Ref.: Hinson, SR et al. (2017). Proc. Natl. Acad. Sci. USA 114(21); 5491-5496).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis : A 1:1,000 dilution from a representative lot detected Aquaporin 4 in rat cerebral cortex tissue.Western Blotting Analysis: A 1:500 dilution from a representative lot detected Aquaporin 4 in human brain tissue lysate.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine, 150 mM NaCl (pH 7.4) with 0.05% sodium azide.
Gene Alias AQP-4;Mercurial-insensitive water channel;MIWC
Gene Symbols Aqp4
Host Species Rabbit
Immunogen GST/HIs-taged recombinant fragment corresponding to 75 amino acids from the C-terminal region of rat Aquaporin-4.
Molecular Weight of Antigen ∽34.0 kDa observed; 34.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001257487.1
Test Specificity This rabbit polyclonal antiody detects Aquaporin-4. It targets an epitope with in 75 amino acids from the C-terminal.
Target Species Rat, Mouse, Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.