Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-5 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Aquaporin-5 |
|---|---|
| Concentration | 1.0mg/mL |
| Dilution | Western Blot 2μg/ml, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 10μg/ml, Immunohistochemistry-Paraffin 1:300 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP267247SS
![]() |
Novus Biologicals
NBP267247SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP267247
![]() |
Novus Biologicals
NBP267247 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Aquaporin-5 Polyclonal specifically detects Aquaporin-5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Aquaporin-5 | |
| Western Blot 2μg/ml, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 10μg/ml, Immunohistochemistry-Paraffin 1:300 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| AQP-5, aquaporin 5, aquaporin-5 | |
| AQP5 | |
| IgG | |
| Affinity Purified |
| 1.0mg/mL | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Cancer, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 362 | |
| Synthetic peptide made to an internal portion of human | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title