Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-5 Antibody (SR1459), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Aquaporin-5 |
---|---|
Clone | SR1459 |
Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Aquaporin-5 Monoclonal antibody specifically detects Aquaporin-5 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
Aquaporin-5 | |
Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AQP-5, aquaporin 5, aquaporin-5 | |
A synthesized peptide derived from human Aquaporin-5 (Uniprot #: P55064) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1459 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
362 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title