Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$192.47 - $517.59
Specifications
| Antigen | Aquaporin-7 |
|---|---|
| Concentration | LYOPH |
| Applications | Immunocytochemistry, Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Aquaporin-7 Polyclonal specifically detects Aquaporin-7 in Human, Mouse, Porcine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| Aquaporin-7 | |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Pig, Equine | |
| AQP-7, AQP7L, AQP9MGC149556, AQPapaquaglyceroporin-7, Aquaglyceroporin-7, aquaporin 7, Aquaporin adipose, aquaporin-7, Aquaporin-7-like, MGC149555 | |
| AQP7 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| O14520 | |
| 364 | |
| IgG | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title