Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aquaporin 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA128171
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA128171 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA128171 Supplier Invitrogen™ Supplier No. PA128171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-28171 detects Aquaporin Adipose Specific from human samples.

Aquaporins/major intrinsic protein (MIP) are a family of water-selective membrane channels. Aquaporin 7 has greater sequence similarity with AQP3 and AQP9 and they may be a subfamily. Aquaporin 7 and AQP3 are at the same chromosomal location suggesting that 9p13 may be a site of an aquaporin cluster. Aquaporin 7 facilitates water, glycerol and urea transport. It may play an important role in sperm function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.09% sodium azide
Gene Aqp7
Gene Accession No. O14520
Gene Alias Aqp7; AQP-7; AQP7L; AQP9; AQPap; Aquaglyceroporin-7; aquaporin 7; aquaporin adipose; Aquaporin-7; Aquaporin-7-like; GLYCQTL; MGC149555; MGC149556; RP11-311H10.6
Gene Symbols Aqp7
Host Species Rabbit
Immunogen Synthetic peptide corresponding to 20aa sequence within the N-terminal, cytoplasmic domain of human Aquaporin, adipose specific (KLH).
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.