Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Aquaporin 8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569362
Description
This target displays homology in the following species: Cow: 86%; Dog: 100%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%; Sheep: 100%.
Aquaporin-8 is a protein that in humans is encoded by the AQP8 gene. Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. Forms a water-specific channel; mercury-sensitive. Not permeable to glycerol or urea. Expressed only in pancreas and colon.
Specifications
Aquaporin 8 | |
Polyclonal | |
Unconjugated | |
Aqp8 | |
AI255744; AQP8; AQP-8; aquaporin 8; Aquaporin8; aquaporin-8; membrane water channel; MGC93492; water channel | |
Rabbit | |
Affinity chromatography | |
RUO | |
29172 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P56405 | |
Aqp8 | |
synthetic peptide directed towards the middle region of rat Aquaporin 8. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction