Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin-9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Aquaporin-9 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Aquaporin-9 Polyclonal specifically detects Aquaporin-9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Aquaporin-9 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
AQP-9, Aquaglyceroporin-9, aquaporin 9, aquaporin-9, HsT17287, Small solute channel 1, SSC1aquaglyceroporin-9 | |
AQP9 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
O43315 | |
366 | |
IgG | |
Unpurified | |
Human. Other species not yet tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title