Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Aquaporin 9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597110 Supplier Invitrogen™ Supplier No. PA597110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGCAMNPARD LSPRLFTALA GWGFEVFRAG NNFWWIPVVG PLVGAVIGGL IYVLVIEIHH PEPDSVFKTE QSEDKPEKYE LSVIM Positive Samples: HL-60, 293T, Mouse liver, Mouse skeletal muscle, Rat liver; Cellular Location: Membrane, Multi-pass membrane protein.

The aquaporins are a family of water-selective membrane channels. This gene encodes a member of a subset of aquaporins called the aquaglyceroporins. This protein allows passage of a broad range of noncharged solutes and also stimulates urea transport and osmotic water permeability. This protein may also facilitate the uptake of glycerol in hepatic tissue. The encoded protein may also play a role in specialized leukocyte functions such as immunological response and bactericidal activity. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Aqp9
Gene Accession No. O43315, Q9JJJ3
Gene Alias 1700020I22Rik; AI266899; AQP9; AQP-9; Aquaglyceroporin-9; aquaporin 9; aquaporin-9; HsT17287; neutral solute channel aquaporin 9; small solute channel 1; SSC1; T17287
Gene Symbols Aqp9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 211-295 of human AQP9 (NP_0661902).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366, 64008
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.