Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin 9 Rabbit anti-Bovine, Mouse, Ovine, Rat, Polyclonal, FabGennix

Catalog No. 502206123
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6123 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6123 Supplier Fabgennix Inc Supplier No. AQP9901AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Aquaporin 9 Polyclonal antibody specifically detects Aquaporin 9 in Bovine, Mouse, Ovine, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Aquaporin 9
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Aqp9
Gene Accession No. P56627, Q9JJJ3
Gene Alias 1700020I22Rik; AI266899; AQP9; AQP-9; Aquaglyceroporin-9; aquaporin 9; aquaporin-9; HsT17287; neutral solute channel aquaporin 9; small solute channel 1; SSC1; T17287
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 250-294 on rat Aquaporin 9 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 443184, 516762, 64008, 65054
Target Species Bovine, Mouse, Ovine, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.