Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AQUASTAR™ Coulomat A (Original Formulation), MilliporeSigma™

Catalog No. max1697a1
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
  • Reagent for Colometric Karl Fischer Titrations.
  • Anolyte containing halogenated hydrocarbons for oily samples.
  • Free of pyridine. For use with all coulometric titrators.
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless to Yellow
Quantity 500 mL
CAS 7553-56-2
Packaging Poly-Coated Amber Glass Bottle
Physical Form Liquid
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
UN Number 1993
Synonym Coulometric Anode Solution
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Chemical Name or Material AQUASTAR Coulomat A, Original Formulation
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid, Toxic
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.