Learn More
AlphaProTech AquaTrak™ Ankle High Boot Cover
Shop All AlphaProTech Products
Click to view available options
Size:
Large
Medium
Universal
X-Large
Description
- Superior Anti-Skid Properties
- Seamless Sole Design
- Low Particle Count
- Sonic Welded Seams
Specifications
Specifications
| Size | Universal |
| Material | Polyethylene, Polypropylene Blend |
| Color | Blue |
| Closure Type | Elastic Top with Ankle Ties |
| Sole Type | Anti-skid |