Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AR-R 17779 hydrochloride, Tocris Bioscience™
SDP

Catalog No. 396410 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-641-0 Supplier R&D Systems Supplier No. 3964/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

α7-selective agonist

Selective agonist of α7 nAChRs (Ki values are 190 and 16000 nM for rat α7 and α4β2 receptors respectively). CNS penetrant after systemic administration.

Chemical Identifiers

CAS 178419-42-6
Molecular Formula C9H15ClN2O2
Molecular Weight (g/mol) 218.681
InChI Key XGLBLUBBDSJBIU-FVGYRXGTSA-N
Synonym ar-r 17779 hydrochloride, ar-r17779 hcl, ar-r17779 hydrochloride, hcl, 2s-4-azaspiro bicyclo 2.2.2 octane-2,2'-1,4 oxazolidin-5'-one hydrochloride, 3s-spiro 1-azabicyclo 2.2.2 octane-3,5'-oxazolidine-2'-one hydrochloride
PubChem CID 9859263
IUPAC Name (5S)-spiro[1,3-oxazolidine-5,3'-1-azabicyclo[2.2.2]octane]-2-one;hydrochloride
SMILES C1CN2CCC1C3(C2)CNC(=O)O3.Cl

Specifications

Quantity 10 mg
Formula Weight 218.68
Percent Purity >98%
Chemical Name or Material AR-R 17779 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.