Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARA24 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985626
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5626 Supplier Bioss Supplier No. BS3751R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).

Specifications

Antigen ARA24
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RAN
Gene Accession No. P62826
Gene Alias AAF30287; Androgen receptor-associated protein 24; ARA24; ARA25; CG1404; CG1404 gene product from transcript CG1404-RC; CG1404-PA; CG1404-PB; CG1404-PC; CG1404-PD; Dmel\CG1404; Dmel_CG1404; dran; Gsp1; GTPase Ran; GTP-binding nuclear protein Ran; guanosine triphosphatase Ran; I79_024397; l(1)G0075; member RAS oncogene family; OK/SW-cl.81; OTTHUMP00000166078; OTTHUMP00000166102; Q9VZ23; ran; Ran GTPase; ran GTP-binding protein; RAN, member RAS oncogene family; RAN, member RAS oncogene family L homeolog; ran.L; ran-1; Ran10A; RanGTP; RanGTPase; Ran-PA; Ran-PB; Ran-PC; Ran-PD; RanTC4; Rasl2-8; Ras-like protein TC4; Ras-related nuclear protein; TC4; XELAEV_18007741mg; xran
Gene Symbols RAN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ARA24.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5901
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.